Specifications
- Antibody Type:Primary
- Antigen Name:Neurotrophin 3
- Antigen Symbol:NT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken,Human,Rat
- Western Blot:Yes
- Size:100 µL
- Epitope:139-149 aa
- Cross Adsorption:No
- Neutralisation:Yes
- Format:Lyophilized
- Antigen Synonyms:NGF-2|HDNF|Ntf3|Neurotrophin-3|Nerve growth factor 2|Ntf-3|Neurotrophic factor
- Immunogen:A synthetic peptide (YAEHKSHRGEY) as part of human (aa: 139-149), mouse and rat NT3 protein conjugated to BSA has been used as the immunogen.
- Cat. No.:10782-386
- Supplier no.:R-097-100
Specifications
About this item
FUNCTION: Seems to promote the survival of visceral and proprioceptive sensory neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family.
A cross reactivity of less than 0.1% to mouse NGF, recombinant human BDNF and NT4/5 has been shown by dot blot.
Application Information:
IHC, ELISA, WB, inhibition of biological activity. A dilution of 1:500 to 1:2000 is recommended for IHC, western blot. For inhibition of biological activity: 1:10-50 for in vitro, 5-10 µl/g body weight for in vivo. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 139-149 aa
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Chicken, Human