Specifications
- Antibody Type:Primary
- Antigen Name:Galectin 3
- Antigen Symbol:GAL3
- Clonality:Monoclonal
- Clone:5C21
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:carbohydrate-binding protein 35|GALIG|galectin-3|IgE-binding protein|GAL3|lectin L-29|CBP35|GALBP|MAC-2 antigen|L31|35 kDa lectin|LGALS2|galactose-specific lectin 3|MAC2|laminin-binding protein
- Immunogen:Full length recombinant Galectin-3 expressed in and purified from E. coli.
- Cat. No.:10782-094
- Supplier no.:M-1574-100
Specifications
About this item
Galectin 3 is a lectin with carbohydrate recognition domains (CRD) which bind -galactoside. It is a multifunctional protein expressed both on the cell surface, cytoplasm and nucleus and appears to have roles in specific carbohydrate binding and in the regulation of mRNA splicing.
Antibody recognizes Galectin 3 by western blots (1:2000-1"10000) and in immunocytochemistry or Immunohistory chemistry on frozen or 4% PFA fixed tissues. Antibody has not been tested with paraffin tissues. Antibody reacts with human, mouse and rat and likely other species because of homology but this has not be confirmed.
Application Information:
WB, ICC. Suggested dilution of at least 1:1,000 for ICC. Suggested dilution of 1:2,000 or lower for WB. Optimal concentrations/dilutions should be determined by the end-user.
Type: Primary
Antigen: Galectin-3
Clonality: Monoclonal
Clone: 5C21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat