Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase isozyme L1
- Antigen Symbol:UCHL1
- Clonality:Monoclonal
- Clone:BH7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:Neuron cytoplasmic protein 9.5|PGP9.5|UCHL1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|Ubiquitin carboxyl-terminal hydrolase isozyme L1
- Immunogen:Recombinant full length human Ubiquitin C Terminal Hydrolase 1 (UCHL1) purified from E. coli.
- Cat. No.:10782-086
- Supplier no.:M-1407-100
Specifications
About this item
This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. The enzyme also binds to free monoubiquitin and may prevent its degradation in lysosomes (ref: SWISSPROT).
The specificity of this antibody has been confirmed by WB. This antibody detects ~24 kDa UCHL1 enzyme. Suitable control tissue is rat spinal cord, brain, SHSY-5Y or HEK293 cell extract.
Application Information:
Western Blotting (WB), Immunocytochemistry (IC) and Immunohistochemistry (IH). A dilution of 1:20,000 is recommended for WB. A dilution of 1:500 - 1:2,000 is recommended for IC and IH. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Ubiquitin C Terminal Hydrolase 1
Clonality: Monoclonal
Clone: BH7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Bovine, Mouse, Pig