Specifications
- Antibody Type:Primary
- Antigen Name:Ghrelin
- Antigen Symbol:GHRL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Guinea pig
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Sheep,Human
- Size:50 µL
- Epitope:amino acids 24-51 from Human ghrelin.
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:ghrelin 28|obestatin|ghrelin 27|Growth hormone-releasing peptide|MTLRP|Protein M46|Motilin-related peptide|Appetite-regulating hormone
- Immunogen:A synthetic peptide (GSSFLSPEHQRVQQRKESKKPPAKLQPR) corresponding to the amino acids 24-51 from human ghrelin. This peptide was conjugated to carrier protein to enhance the immunological response.
- Cat. No.:10782-014
- Supplier no.:GP-032-50
Specifications
About this item
Ghrelin is the ligand for growth hormone secretagogue receptor type 1 (GHSR) and upon binding to the receptor it induces the release of growth hormone from the pituitary. This ligand has an appetite-stimulating effect and is involved in growth regulation.
Specificity was demonstrated by immunohistochemistry.
Application Information:
IHC. A concentration of 1 in 3000 is recommended for IHC. IHC performed in sheep abomasum demonstrates intense staining of cells and terminals. No staining is evident when the primary antibody is pre-absorbed with 0.5 mg/ml of human ghrelin. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ghrelin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 24-51 from Human ghrelin.
Host: Guinea Pig
Isotype: IgG
Reactivity: Sheep, Human