- Description:MAS-100™ Eco Airsampler for Food Industry
- Environmentally Preferable:
- Cat. No.:EM1.09227.0001
- Supplier no.:1.09227.0001
MAS-100 Eco® is an economical, small, and lightweight microbial air sampler.
It is compatible with Petri dishes and offers precise, reproducible measurements, excellent collection efficiency, built-in airflow compensation as well as fully automatic calibration. Both hardware and software are validated for HACCP and validation documents are in accordance with ISO14698 parts one and two.