Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant PDGF-AA (from E. coli)
 
undefined
Human Recombinant PDGF-AA (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: NR6R-3T3 cell proliferation, ED50 ≤50 ng/ml, bioactivity ≥2.0×10⁴ units/mg
  • Gene ID:
    P04085
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    ODGF|GDGF
  • UniProtKB:
    P04085
  • Protein/Peptide Name:
    PDGF-AA
  • Purity:
    ≥95%
  • Molecular Weight:
    14.4/28.9 (kDa)
  • Sequence:
    MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

 

Frequently Bought Together