Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IFNg (from E. coli)
Human Recombinant IFNg (from E. coli)
Certificates
undefined
Human Recombinant IFNg (from E. coli)
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0.75 ng/ml, Bioactivity ≥1.3×10⁶ units/mg
  • Gene ID:
    P01579
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    Type II interferon|T cell interferon|MAF|Immune Interferon
  • Protein/Peptide Name:
    IFNg
  • Purity:
    ≥95%
  • Molecular Weight:
    16.9 kDa
  • Sequence:
    MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together