Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant IGF-II (from <i>E. coli</i>)

Human Recombinant IGF-II (from E. coli)

Human Recombinant IGF-II (from <i>E. coli</i>)
Certificates
Human Recombinant IGF-II (from <i>E. coli</i>)

5 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.

  • High quality
  • Low endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10787-306
  • 10787-310
  • 10822-062
  • 10821-832
  • 10787-308
  • Protein/Peptide Name:
  • IGF-II
  • IGF-II
  • IGF-II
  • IGF-II
  • IGF-II
  • Protein Synonyms:
  • Somatomedin A
  • Somatomedin A
  • Somatomedin A
  • Somatomedin A
  • Somatomedin A
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P01344
  • P01344
  • P01344
  • P01344
  • P01344
  • Sequence:
  • AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
  • AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
  • AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
  • AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
  • AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-Free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-Free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin Level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin Content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • UniProtKB:
  • P01344
  • P01344
  • P01344
  • P01344
  • P01344
  • Biological Activity:
  • Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
  • Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
  • Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
  • Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
  • Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • Molecular Weight:
  • 7.5 kDa
  • 7.5 kDa
  • 7.5 kDa
  • 7.5 kDa
  • 7.5 kDa
  • Reconstitution Instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage Conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping Temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested Applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Origin
    Availability
    Price
  • Vial
    10 µg
    US
    Log in for availability
    Log in for availability
    Each (10µG)
    Each (10µG) - $92.72
    $92.72
    Each (10µG)
       $92.72
    Minimum order quantity is 1

    Supplier Number: 100-14-10UG
    Specifications:
    More
    Specifications
    Cat. No.
    10787-306
    Protein/Peptide Name
    IGF-II
    Protein Synonyms
    Somatomedin A
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01344
    Sequence
    AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P01344
    Biological Activity
    Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    7.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    50 µg
    US
    Log in for availability
    Log in for availability
    Each (50µG)
    Each (50µG) - $213.21
    $213.21
    Each (50µG)
       $213.21
    Minimum order quantity is 1

    Supplier Number: 100-14-50UG
    Specifications:
    More
    Specifications
    Cat. No.
    10787-310
    Protein/Peptide Name
    IGF-II
    Protein Synonyms
    Somatomedin A
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01344
    Sequence
    AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P01344
    Biological Activity
    Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    7.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    100 µg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $405.19
    $405.19
    Each (100µG)
       $405.19
    Minimum order quantity is 1

    Supplier Number: 100-14-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10822-062
    Protein/Peptide Name
    IGF-II
    Protein Synonyms
    Somatomedin A
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01344
    Sequence
    AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P01344
    Biological Activity
    Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    7.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    500 µg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $941.69
    $941.69
    Each (500µG)
       $941.69
    Minimum order quantity is 1

    Supplier Number: 100-14-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10821-832
    Protein/Peptide Name
    IGF-II
    Protein Synonyms
    Somatomedin A
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01344
    Sequence
    AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P01344
    Biological Activity
    Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    7.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $1,701.07
    $1,701.07
    Each (1mg)
       $1,701.07
    Minimum order quantity is 1

    Supplier Number: 100-14-1MG
    Specifications:
    More
    Specifications
    Cat. No.
    10787-308
    Protein/Peptide Name
    IGF-II
    Protein Synonyms
    Somatomedin A
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01344
    Sequence
    AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P01344
    Biological Activity
    Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    7.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.