Macrophage inflammatory protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta, and bone marrow. MIP-4 receptors include the chemokine receptor 8 (CCR8), the G protein-coupled receptor 30 (GPR30), and the phosphatidylinositol transfer protein membrane-associate 3 (PITPNM3). MIP-4 acts as a chemoattractant for naive T cells, CD4+ T cells, CD8+ T cells, and nonactivated lymphocytes. Further, MIP-4 promotes breast cancer metastasis and attenuates the activation of acute lymphocytic leukemia B cells.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P55774
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:DC-CK1|CCL18|AMAC-1|PARC
- Protein/Peptide Name:MIP-4
- Purity:≥95%
- Molecular Weight:7.9 kDa
- Sequence:AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT