Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant MIP-3 B (from E. coli)
 
undefined
Human Recombinant MIP-3 B (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q99731
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    CCL19|SCYA19|ELC|exodus-3
  • UniProtKB:
    Q99731
  • Protein/Peptide Name:
    MIP-3 B
  • Purity:
    ≥95%
  • Molecular Weight:
    8.8 kDa
  • Sequence:
    GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

 

Frequently Bought Together