Anti-UCHL1 Mouse Monoclonal Antibody [clone: SPM574]
Catalog # 10743-726
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen Symbol:UCHL1
- Clonality:Monoclonal
- Clone:SPM574
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7345
- Antigen Synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Storage Buffer:PBS (0.05% BSA, 0.05% sodium azide).
- Storage Temperature:Store at 4°C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human UCHL1 from brain.
- Purification:Protein A/G purification
- Cat. No.:10743-726
- Supplier no.:MAB12063
Specifications
About this item
Mouse monoclonal antibody raised against native UCHL1.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: SPM574
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: