To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Granulocyte
- Antigen Symbol:Granulocyte
- Clonality:Monoclonal
- Clone:SPM250
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS (0.05% BSA, 0.05% sodium azide).
- Storage Temperature:Store at 4°C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
- Purification:Protein A/G purification
- Cat. No.:10716-368
- Supplier no.:MAB12080
Mouse monoclonal antibody raised against Granulocyte.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Granulocyte
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: SPM250
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: