To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:FK506 Binding Protein 4
- Antigen Symbol:FKBP4
- Clonality:Monoclonal
- Clone:Hi52C
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2288
- Antigen Synonyms:p52|FKBP52|Hsp56|FKBP59|FKBP51|HBI|PPIase
- Storage Buffer:PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to residues of human FKBP4.
- Purification:Protein G purification
- Cat. No.:10714-790
- Supplier no.:MAB11372
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of FKBP4.
Recommended Dilutions: Western Blot: 1:2000; Immunohistochemistry: 1:250; Immunoprecipitation: 5 ?g with 10-20 ?l Protein A beads
Type: Primary
Antigen: FKBP4
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: Hi52C
Epitope:
Host: IgG
Isotype: Mouse
Reactivity: Dog, Hamster, Human, Mouse, Rat