Anti-ADRB2 Rabbit Polyclonal Antibody
Catalog # 10683-204
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Adrenoceptor Beta 2
- Antigen Symbol:ADRB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Monkey,Gorilla,Chimpanzee,Human
- Size:50 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:154
- Antigen Synonyms:BETA2AR|ADRBR|Beta-2 adrenoreceptor|BAR|ADRB2R|Beta-2 adrenoceptor|B2AR
- Storage Buffer:PBS (0.1% sodium azide)
- Storage Temperature:Store at 4°C. For long term storage store at –80 °C . Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids at 5th transmembrane domain of human ADRB2.
- Purification:Immunoaffinity chromatography
- Cat. No.:10683-204
- Supplier no.:PAB25558
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ADRB2.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 17-26 ?g/ml
Type: Primary
Antigen: ADRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human, Monkey