To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Dickopf WNT Signaling Pathway Inhibitor 3
- Antigen Symbol:DKK3
- Clonality:Monoclonal
- Clone:4G7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:27122
- Antigen Synonyms:RIG|REIC
- Storage Buffer:0.03% sodium azide
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human DKK3.
- Cat. No.:10710-786
- Supplier no.:MAB10625
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant DKK3.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: DKK3
Clonality: Monoclonal
Clone: 4G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey