Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-DNMBP Rabbit Polyclonal Antibody
Anti-DNMBP Rabbit Polyclonal Antibody
Catalog # 10583-932
Supplier:  Abnova
undefined
Anti-DNMBP Rabbit Polyclonal Antibody
Catalog # 10583-932
Supplier:  Abnova
Supplier Number:  H00023268-D01

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Dynamin Binding Protein
  • Antigen Symbol:
    DNMBP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    23268
  • Antigen Synonyms:
    TUBA|ARHGEF36
  • Amino Acid Number:
    1 to 823
  • Storage Buffer:
    No additive
  • Storage Temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    DNMBP (AAH41628.1, 1 a.a. ~ 823 a.a) full-length human protein.
  • Cat. No.:
    10583-932
  • Supplier no.:
    H00023268-D01

Specifications

About this item

Rabbit polyclonal antibody raised against a full-length human DNMBP protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: DNMBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human