Anti-PSMD10 Mouse Monoclonal Antibody [clone: 1B4]
Catalog # 10598-366
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome (prosome macropain) 26S Subunit non-ATPase 10
- Antigen Symbol:PSMD10
- Clonality:Monoclonal
- Clone:1B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5716
- Antigen Synonyms:dJ889N15.2|p28(GANK)|p28
- Amino Acid Number:127 to 226
- Storage Buffer:1x PBS, pH 7.4
- Sequence:EGGANPDAKDHYEATAMHRAAAKGNLKMIHILLYYKASTNIQDTEGNTPLHLACDEERVEEAKLLVSQGASIYIENKEEKTPLQVAKGGLGLILKRMVEG
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:PSMD10 (AAH11960, 127 a.a. ~ 226 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Cat. No.:10598-366
- Supplier no.:H00005716-M22
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant PSMD10.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: PSMD10
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human