Anti-PSMC6 Mouse Monoclonal Antibody [clone: 1E5]
Catalog # 10598-362
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome (prosome macropain) 26S Subunit ATPase 6
- Antigen Symbol:PSMC6
- Clonality:Monoclonal
- Clone:1E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5706
- Antigen Synonyms:SUG2|P44|p42|CADP44|HEL-S-73
- Amino Acid Number:1 to 389
- Storage Buffer:1x PBS, pH 7.4
- Sequence:MADPRDKALQDYRKKLLEHEEIDGRLKELREQLKELTKQYEKSENDLKALQSVGQIVGEVLKQLTEEKFIVKATNGPRYVVGCRRQLDKSKLKPGTRVALDMTTLTIMRYLPREVDPLVYNMSHEDPGNVSYSEIGGLSEQIRELREVIELPLTNPELFQRVGIIPPKGCLLYGPPGTGKTLLARAVASQLDCNFLKVVSSSIVDKYIGESARLIREMFNYARDHQPCIIFMGEIDAIGGRRFSEGTSADREIQRTLMELLNQMDGFDTLHRVKMIMATNRPDTLDPALLRPGRLDRKIHIDLPNEQARLDILKIHAGPITKHGEIDYEAIVKLSDGFNGADLRNVCTEAGMFAIRADHDFVVQEDFMKAVRKVADSKKLESKLDYKPV
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:PSMC6 (AAH05390, 1 a.a. ~ 389 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Cat. No.:10598-362
- Supplier no.:H00005706-M04
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant PSMC6.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: PSMC6
Clonality: Monoclonal
Clone: 100000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human