Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MAP3K4 Mouse Monoclonal Antibody [clone: X1]
Catalog # 10625-206
Supplier:  Abnova
undefined
Anti-MAP3K4 Mouse Monoclonal Antibody [clone: X1]
Catalog # 10625-206
Supplier:  Abnova
Supplier Number:  H00004216-M11A

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Mitogen-activated Protein Kinase Kinase Kinase 4
  • Antigen Symbol:
    MAP3K4
  • Clonality:
    Monoclonal
  • Clone:
    X1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    200 µL
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites fluid
  • Gene ID:
    4216
  • Antigen Synonyms:
    MTK1|MEKK4|PRO0412|MEKK 4|MAPKKK4
  • Amino Acid Number:
    1201 to 1300
  • Sequence:
    AASRPSPSGGDSVLPKSISSAHDTRGSSVPENDRLASIAAELQFRSLSRHSSPTEERDEPAYPRGDSSGSTRRSWELRTLISQSKDTASKLGPIEAIQKS
  • Storage Temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    MAP3K4 (NP_005913, 1201 a.a. ~ 1300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Cat. No.:
    10625-206
  • Supplier no.:
    H00004216-M11A

Specifications

About this item

Mouse monoclonal antibody raised against a partial recombinant MAP3K4.

Type: Primary
Antigen: MAP3K4
Clonality: Monoclonal
Clone: X1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse