Anti-HOXA4 Mouse Polyclonal Antibody
Catalog # 10558-550
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Homeobox A4
- Antigen Symbol:HOXA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:3201
- Antigen Synonyms:HOX1|HOX1D
- Amino Acid Number:1 to 320
- Storage Buffer:1x PBS, pH 7.4
- Sequence:MTMSSFLINSNYIEPKFPPFEEYAQHSGSGGADGGPGGGPGYQQPPAPPTQHLPLQQPQLPHAGGGREPTASYYAPRTAREPAYPAAALYPAHGAADTAYPYGYRGGASPGRPPQPEQPPAQAKGPAHGLHASHVLQPQLPPPLQPRAVPPAAPRRCEAAPATPGVPAGGSAPACPLLLADKSPLGLKGKEPVVYPWMKKIHVSAVNPSYNGGEPKRSRTAYTRQQVLELEKEFHFNRYLTRRRRIEIAHTLCLSERQVKIWFQNRRMKWKKDHKLPNTKMRSSNSASASAGPPGKAQTQSPHLHPHPHPSTSTPVPSSI
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:HOXA4 (AAI60036.1, 1 a.a. ~ 320 a.a) full-length human protein.
- Cat. No.:10558-550
- Supplier no.:H00003201-B01P
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human HOXA4 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: HOXA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human