Anti-VIM Mouse Monoclonal Antibody [clone: 4F2E9]
10744-204
: Abnova
- Antibody Type:Primary
- Antigen Name:Vimentin
- Antigen Symbol:VIM
- Clonality:Monoclonal
- Clone:4F2E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:7431
- Antigen Synonyms:CTRCT30|HEL113
- Amino Acid Number:2 to 466
- Storage Buffer:0.03% sodium azide
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 2-466 of human VIM.
- Cat. No.:10744-204
- Supplier no.:MAB10419
Mouse monoclonal antibody raised against partial recombinant VIM.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 4F2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human