Anti-CYP26A1 Mouse Monoclonal Antibody [clone: F27 P6 A1]
10676-116
: Abnova
- Antibody Type:Primary
- Antigen Name:Cytochrome P450, Family 26, Subfamily A, Polypeptide 1
- Antigen Symbol:CYP26A1
- Clonality:Monoclonal
- Clone:F27 P6 A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:200 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:1592
- Antigen Synonyms:CP26|P450RAI|P450RAI1|CYP26
- Storage Buffer:PBS (0.08% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of human CYP26A1.
- Cat. No.:10676-116
- Supplier no.:MAB1857
Mouse monoclonal antibody raised against synthetic peptide of CYP26A1.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-5 ?g/ml
Type: Primary
Antigen: CYP26A1
Clonality: Monoclonal
Clone: F27 P6 A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: