Order Entry
United States
ContactUsLinkComponent
 
Anti-MAP1A Mouse Monoclonal Antibody [clone: MP-1]
  10724-614
 :  Abnova
undefined
Anti-MAP1A Mouse Monoclonal Antibody [clone: MP-1]
  10724-614
 :  Abnova
 :  MAB1520
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Microtubule-associated protein 1A
  • Antigen Symbol:
    MAP1A
  • Clonality:
    Monoclonal
  • Clone:
    MP-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rat
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Lyophilized
  • Gene ID:
    25152
  • Antigen Synonyms:
    MAP1a|MTAP-1|6330416M19Rik|MTAP1|MOTH1|AI853608
  • Storage Buffer:
    1.2% sodium acetate (2 mg BSA, 0.01 mg sodium azide)
  • Storage Temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified rat Map1a.
  • Purification:
    Affinity purification
  • Cat. No.:
    10724-614
  • Supplier no.:
    MAB1520

 

 

Mouse monoclonal antibody raised against native Map1a.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml; Immunohistochemistry (Frozen sections): 1-2 ?g/ml

Type: Primary
Antigen: MAP1A
Clonality: Monoclonal
Clone: MP-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat