Anti-TNN Mouse Monoclonal Antibody [clone: T20]
Catalog # 10742-278
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Troponin 1
- Antigen Symbol:TNN1
- Clonality:Monoclonal
- Clone:T20
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:63923
- Storage Buffer:1.2% sodium acetate (2 mg BSA, 0.01 mg sodium azide)
- Storage Temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human TNN.
- Purification:Affinity purification
- Cat. No.:10742-278
- Supplier no.:MAB1529
Specifications
About this item
Mouse monoclonal antibody raised against native TNN.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml; Immunohistochemistry (Frozen sections): 2-4 ?g/ml
Type: Primary
Antigen: TNN
Clonality: Monoclonal
Clone: T20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human