Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 2B7]
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # 10730-122
Supplier:  Abnova
undefined
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # 10730-122
Supplier:  Abnova
Supplier Number:  MAB10302

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Protein Kinase AMP-activated Alpha 1 Catalytic Subunit
  • Antigen Symbol:
    PRKAA1
  • Clonality:
    Monoclonal
  • Clone:
    2B7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    5562
  • Antigen Synonyms:
    catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
  • Storage Buffer:
    0.03% sodium azide
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human PRKAA1.
  • Cat. No.:
    10730-122
  • Supplier no.:
    MAB10302

Specifications

About this item

Mouse monoclonal antibody raised against recombinant PRKAA1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: PRKAA1
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat