Anti-TAAR1 Rabbit Polyclonal Antibody
Catalog # 10740-396
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Trace amine-associated Receptor 1
- Antigen Symbol:TAAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Chimpanzee,Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:134864
- Antigen Synonyms:TRAR1|Trace amine receptor 1|TaR-1|TA1|TAR1
- Amino Acid Number:200 to 230
- Storage Buffer:PBS (0.05% BSA, 0.05% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 200-230 of human TAAR1.
- Cat. No.:10740-396
- Supplier no.:PAB0313
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant TAAR1.
Type: Primary
Antigen: TAAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human