Anti-ANKRD26 Rabbit Polyclonal Antibody
Catalog # 10689-142
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Ankyrin Repeat Domain 26
- Antigen Symbol:ANKRD26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:232339
- Antigen Synonyms:BC008111|mKIAA1074|5730521P14Rik
- Amino Acid Number:1 to 15
- Storage Buffer:20 mM KH2PO4, 150 mM NaCl, pH 7.2 (0.01% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1-15 of mouse Ankrd26.
- Cat. No.:10689-142
- Supplier no.:PAB10090
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Ankrd26.
Recommended Dilutions: ELISA: 1:20000-1:80000; Western Blot: 1:500-1:3000
Type: Primary
Antigen: ANKRD26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat