Anti-MBD1 Rabbit Polyclonal Antibody
Catalog # 10725-048
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl-CpG Binding Domain Protein 1
- Antigen Symbol:MBD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:4152
- Antigen Synonyms:CXXC3|PCM1|RFT
- Amino Acid Number:98 to 113 and 391 to 405
- Storage Buffer:PBS (0.02% sodium azide)
- Sequence:RQVGPQSGEVRKEAPR, SESEDGAGSPPPYRR
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A mixture of synthetic peptides corresponding to amino acids 98-113 and 391-405 of human MBD1.
- Cat. No.:10725-048
- Supplier no.:PAB0172
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MBD1.
Type: Primary
Antigen: MBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human