Anti-H2-D2 Mouse Monoclonal Antibody [clone: 27-11-13S]
10735-006
: Abnova
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:H2-D2
- Clonality:Monoclonal
- Clone:27-11-13S
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Size:500 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:100 mM BBS, pH 8.2
- Storage Temperature:Store at 4°C.
- Immunogen:Native from C3H alphaBDF1 splenocytes.
- Cat. No.:10735-006
- Supplier no.:MAB3261
Mouse monoclonal antibody raised against H2-D2.
Type: Primary
Antigen: H2-D2
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 27-11-13S
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Mouse