Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-H-2KK Mouse Monoclonal Antibody [clone: 16-3-22S]
Anti-H-2KK Mouse Monoclonal Antibody [clone: 16-3-22S]
Catalog # 10735-036
Supplier:  Abnova
Anti-H-2KK Mouse Monoclonal Antibody [clone: 16-3-22S]
Catalog # 10735-036
Supplier:  Abnova
Supplier Number:  MAB3263

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    MHC Class I (H-2Kk)
  • Antigen Symbol:
    H-2KK
  • Clonality:
    Monoclonal
  • Clone:
    16-3-22S
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Size:
    500 µg
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Antigen Synonyms:
    H2K1|H2K|H2KK|H-2 class I histocompatibility antigen K-K alpha chain
  • Storage Buffer:
    100 mM BBS, pH 8.2
  • Storage Temperature:
    Store at 4°C.
  • Immunogen:
    C3H.SW mouse splenocytes.
  • Cat. No.:
    10735-036
  • Supplier no.:
    MAB3263

Specifications

About this item

Mouse monoclonal antibody raised against H-2Kk.

Type: Primary
Antigen: H-2Kk
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 16-3-22S
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Mouse