Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PRKAA1 Rabbit Polyclonal Antibody
Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # 10675-148
Supplier:  Abnova
undefined
Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # 10675-148
Supplier:  Abnova
Supplier Number:  PAB12747

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Protein Kinase AMP-activated Alpha 1 Catalytic Subunit
  • Antigen Symbol:
    PRKAA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Fish,
    Insect,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Antigen Synonyms:
    catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
  • Modification:
    phospho-Ser79
  • Storage Buffer:
    TBS, pH 7.2 (10% Proclin300)
  • Sequence:
    CGSPN
  • Storage Temperature:
    Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide corresponding to residues surrounding S79 of Carassius auratus PRKAA1.
  • Cat. No.:
    10675-148
  • Supplier no.:
    PAB12747

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKAA1.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml

Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Fish, Human, Insect, Mouse, Rat