Anti-EME1 Mouse Monoclonal Antibody [clone: 7h2/1]
10683-914
: Abnova
- Antibody Type:Primary
- Antigen Name:essential meiotic structure-specific endonuclease 1
- Antigen Symbol:EME1
- Clonality:Monoclonal
- Clone:7h2/1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:146956
- Antigen Synonyms:MMS4L|SLX2A
- Storage Buffer:PBS (0.1% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human EME1.
- Cat. No.:10683-914
- Supplier no.:MAB2490
Mouse monoclonal antibody raised against EME1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: EME1
Clonality: Monoclonal
Clone: 7h2/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human