Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CENPU Rabbit Polyclonal Antibody
Anti-CENPU Rabbit Polyclonal Antibody
  10732-742
 :  Abnova
undefined
Anti-CENPU Rabbit Polyclonal Antibody
  10732-742
 :  Abnova
 :  PAB21473
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Centromere protein U
  • Antigen Symbol:
    CENPU
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    79682
  • Antigen Synonyms:
    MLF1IP|CENPU50|PBIP1|KLIP1|CENP50
  • Storage Buffer:
    PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
  • Sequence:
    EETYETFDPPLHSTAIYADEEEFSKHCGLSLSSTPPGKEAKRSSDTSGNEASEIESVKISAKKPGRKLRPISDDSESIEESDTRRKVKSAEKISTQRHEVIRTTASSELSEKPAESV
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human MLF1IP.
  • Purification:
    Antigen affinity purification
  • Cat. No.:
    10732-742
  • Supplier no.:
    PAB21473

 

 

Rabbit polyclonal antibody raised against recombinant MLF1IP.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: CENPU
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: