Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # 10675-150
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase AMP-activated Alpha 1 Catalytic Subunit
- Antigen Symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Insect,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5562
- Antigen Synonyms:catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
- Modification:phospho-Thr172
- Storage Buffer:TBS, pH 7.2 (10% Proclin300)
- Sequence:LRTS
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding T172 of human PRKAA1.
- Cat. No.:10675-150
- Supplier no.:PAB12602
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKAA1.
Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Human, Insect, Mouse, Rat