To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:FK506 Binding Protein 4
- Antigen Symbol:FKBP4
- Clonality:Monoclonal
- Clone:AT4D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2288
- Antigen Synonyms:p52|FKBP52|Hsp56|FKBP59|FKBP51|HBI|PPIase
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human FKBP4.
- Cat. No.:10714-788
- Supplier no.:MAB3587
Mouse monoclonal antibody raised against full length recombinant FKBP4.
Type: Primary
Antigen: FKBP4
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: AT4D3
Epitope:
Host: IgG2b kappa
Isotype: Mouse
Reactivity: Human