Anti-PSMD10 Rabbit Polyclonal Antibody
Catalog # 10730-430
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome (prosome macropain) 26S Subunit non-ATPase 10
- Antigen Symbol:PSMD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Pig,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5716
- Antigen Synonyms:dJ889N15.2|p28(GANK)|p28
- Amino Acid Number:100 to 200
- Storage Buffer:Tris-glycine, 150 mM NaCl (0.05% sodium azide)
- Storage Temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 100-200 of human PSMD10.
- Cat. No.:10730-430
- Supplier no.:PAB12056
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of PSMD10.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type: Primary
Antigen: PSMD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat, Zebrafish