Anti-INS Mouse Monoclonal Antibody [clone: 2/5E4]
Catalog # 10627-842
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Insulin
- Antigen Symbol:INS
- Clonality:Monoclonal
- Clone:2/5E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:1 mg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:3630
- Antigen Synonyms:MODY10|IDDM|ILPR|IDDM2|IRDN|IDDM1
- Storage Buffer:BBS
- Storage Temperature:Store at 4°C.
- Immunogen:Recombinant protein corresponding to human INS.
- Cat. No.:10627-842
- Supplier no.:MAB3684
Specifications
About this item
Mouse monoclonal antibody raised against recombinant INS.
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: 2/5E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human