Anti-CLEC4E Mouse Monoclonal Antibody [clone: AT16E3]
10708-160
: Abnova
- Antibody Type:Primary
- Antigen Name:C-type lectin Domain family 4, member E
- Antigen Symbol:CLEC4E
- Clonality:Monoclonal
- Clone:AT16E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:26253
- Antigen Synonyms:MINCLE|CLECSF9
- Amino Acid Number:41 to 219
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 41-219 of human CLEC4E.
- Cat. No.:10708-160
- Supplier no.:MAB8479
Mouse monoclonal antibody raised against partial recombinant CLEC4E.
Recommended Dilutions: ELISA: 1:250; Western Blot: 1:250-1:500; Immunofluorescence: 1:250-1:500
Type: Primary
Antigen: CLEC4E
Clonality: Monoclonal
Clone: AT16E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: