Anti-PSMD11 Mouse Monoclonal Antibody [clone: AT1F4]
Catalog # 10730-434
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome (prosome macropain) 26S Subunit non-ATPase 11
- Antigen Symbol:PSMD11
- Clonality:Monoclonal
- Clone:AT1F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5717
- Antigen Synonyms:26S proteasome regulatory subunit S9|PSMD11|26S proteasome regulatory subunit p44.5|26S proteasome non-ATPase regulatory subunit 11
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human PSMD11.
- Cat. No.:10730-434
- Supplier no.:MAB5643
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant PSMD11.
Type: Primary
Antigen: PSMD11
Clonality: Monoclonal
Clone: AT1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse