Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ADAMTS4 Rabbit Polyclonal Antibody
Anti-ADAMTS4 Rabbit Polyclonal Antibody
  10683-148
 :  Abnova
undefined
Anti-ADAMTS4 Rabbit Polyclonal Antibody
  10683-148
 :  Abnova
 :  PAB16218

 

  • Antibody Type:
    Primary
  • Antigen Name:
    ADAM Metallopeptidase With Thrombospondin Type 1 Motif, 4
  • Antigen Symbol:
    ADAMTS4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Horse,
    Monkey,
    Orangutan,
    Human,
    Dog
  • Size:
    50 µg
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    9507
  • Antigen Synonyms:
    Aggrecanase 1|ADAM-TS 4|ADMP-1|A disintegrin and metalloproteinase with thrombospondin motifs 4|ADAM-TS4
  • Storage Buffer:
    PBS (0.1% sodium azide)
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to human ADAMTS4.
  • Purification:
    Immunoaffinity purification
  • Cat. No.:
    10683-148
  • Supplier no.:
    PAB16218

 

 

Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS4.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.6 ?g/ml

Type: Primary
Antigen: ADAMTS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Horse, Human, Monkey, Orangutan