Anti-PRKAA1 Rabbit Polyclonal Antibody
10730-124
: Abnova
- Antibody Type:Primary
- Antigen Name:Protein Kinase AMP-activated Alpha 1 Catalytic Subunit
- Antigen Symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Bovine,Rat
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5562
- Antigen Synonyms:catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
- Storage Buffer:Tris-citrate/phosphate, pH 7-8 (0.1% sodium azide)
- Storage Temperature:Store at 4°C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to the portion encoded within exon 8 of human PRKAA1.
- Cat. No.:10730-124
- Supplier no.:PAB14475
Rabbit polyclonal antibody raised against synthetic peptide of PRKAA1.
Recommended Dilutions: Western Blot: 0.2-2 ?g/ml; Immunoprecipitation: 2-8 ?g/mg)
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Rat