Anti-SLC7A5 Rabbit Polyclonal Antibody
Catalog # 10692-112
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 7 Member 5
- Antigen Symbol:SLC7A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Size:50 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:8140
- Antigen Synonyms:E16|SLC7A5|y+ system)|CD98 light chain|4F2LC|hLAT1|large neutral amino|LAT1|CD98|MPE16|D16S469E|solute carrier family 7 (cationic amino acid transporter|member 5|4F2 light chain|integral membrane protein E16|L-type amino acid transporter 1
- Storage Buffer:PBS (0.1% sodium azide)
- Storage Temperature:Store at 4°C. For long term storage store at –80 °C . Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to human SLC7A5.
- Purification:Immunoaffinity purification
- Cat. No.:10692-112
- Supplier no.:PAB16324
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SLC7A5.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 13-26 ?g/ml
Type: Primary
Antigen: SLC7A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human