Anti-ICMT Rabbit Polyclonal Antibody
10678-958
: Abnova
- Antibody Type:Primary
- Antigen Name:IsoprenylCysteine carboxyl methyltransferase
- Antigen Symbol:ICMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:23463
- Antigen Synonyms:PCCMT|Prenylated protein carboxyl methyltransferase|pcCMT|Isoprenylcysteine carboxylmethyltransferase|Prenylcysteine carboxyl methyltransferase|PPMT|Protein-S-isoprenylcysteine O-methyltransferase|ICMT
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ICMT.
- Purification:Protein G purification
- Cat. No.:10678-958
- Supplier no.:PAB2513
Rabbit polyclonal antibody raised against synthetic peptide of ICMT.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: ICMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human