Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MBD6 Rabbit Polyclonal Antibody
  10725-064
 :  Abnova
undefined
Anti-MBD6 Rabbit Polyclonal Antibody
  10725-064
 :  Abnova
 :  PAB23369

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Methyl-CpG Binding Domain 6 DNA Glycosylase
  • Antigen Symbol:
    MBD6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    114785
  • Storage Buffer:
    PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
  • Sequence:
    MERSPRRTHHWQHNGELAEGGAEPKDPPPPGPHSEDLKVPPGVVRKSRRGRRRKYNPTRNSNSSRQDITLEPSPTARAA
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human MBD6.
  • Purification:
    Antigen affinity purification
  • Cat. No.:
    10725-064
  • Supplier no.:
    PAB23369

 

 

Rabbit polyclonal antibody raised against recombinant MBD6.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: MBD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human