Anti-Myeloid Specific Antigen Mouse Monoclonal Antibody [clone: BM-2]
Catalog # 10726-334
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Myeloid Specific Antigen
- Antigen Symbol:Myeloid Specific Antigen
- Clonality:Monoclonal
- Clone:BM-2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS (0.05% sodium azide)
- Storage Temperature:Store at 4°C.
- Immunogen:Native purified from pokeweed mitogen stimulated human peripheral blood lymphocyte nuclei.
- Cat. No.:10726-334
- Supplier no.:MAB5183
Specifications
About this item
Mouse monoclonal antibody raised against native Myeloid Specific Antigen.
Type: Primary
Antigen: Myeloid Specific Antigen
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: BM-2
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human