Anti-UCHL3 Rabbit Polyclonal Antibody
10686-496
: Abnova
- Antibody Type:Primary
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase isozyme L3
- Antigen Symbol:UCHL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gorilla,Chicken,Human,Dog,Rat,Mouse
- Size:50 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7347
- Antigen Synonyms:Ubiquitin thioesterase L3|Ubiquitin thiolesterase L3|UCH L3|UCH-L3|UCHL3_HUMAN.|Ubiquitin thiolesterase
- Storage Buffer:PBS (0.1% sodium azide)
- Storage Temperature:Store at 4°C. For long term storage store at –80 °C . Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to human UCHL3.
- Purification:Immunoaffinity purification
- Cat. No.:10686-496
- Supplier no.:PAB16473
Rabbit polyclonal antibody raised against synthetic peptide of UCHL3.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 ?g/ml
Type: Primary
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Dog, Gorilla, Human, Mouse, Rat