Anti-CREB1 Mouse Monoclonal Antibody [clone: A3A4]
Catalog # 10735-854
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:cAMP responsive element binding protein 1
- Antigen Symbol:CREB1
- Clonality:Monoclonal
- Clone:A3A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:500 µg
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:1385
- Antigen Synonyms:CREB
- Modification:phospho-Ser133
- Storage Buffer:10 mM PBS, pH 7.0
- Storage Temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S133 of human CREB1.
- Cat. No.:10735-854
- Supplier no.:MAB6422
Specifications
About this item
Mouse monoclonal antibody raised against synthetic phosphopeptide of CREB1.
Recommended Dilutions: ELISA: 1 ?g/ml
Type: Primary
Antigen: CREB1
Clonality: Monoclonal
Clone: A3A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: