Anti-MBD1 Rabbit Polyclonal Antibody
Catalog # 10685-288
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl-CpG Binding Domain Protein 1
- Antigen Symbol:MBD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:4152
- Antigen Synonyms:CXXC3|PCM1|RFT
- Storage Buffer:PBS (0.05% sodium azide, 0.05% procl300)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A mixture of synthetic peptides (conjugated with KLH) that two corresponding to C-terminus and one to N-terminus of human MBD1.
- Cat. No.:10685-288
- Supplier no.:PAB14113
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MBD1.
Recommended Dilutions: ELISA: 1:500-1:1000; Western Blot: 1:2000
Type: Primary
Antigen: MBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human