Anti-PRKAG1 Rabbit Polyclonal Antibody
10679-774
: Abnova
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 1
- Antigen Symbol:PRKAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5571
- Antigen Synonyms:5'-AMP-activated protein kinase subunit gamma-1|AMPKG3|AMPK gamma3|PRKAG3|AMPK- gamma3|5'-AMP-activated protein kinase subunit gamma-2|AMPK- gamma1|AMPK subunit gamma-1|AMPKg|AMPK subunit gamma-3|H91620p|PRKAG2|AMPK- gamma2|AMPK subunit gamma-2|PRKAG1|AMPK gamma2|AMPK gamma1|5'-AMP-activated protein kinase subunit gamma-3
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to internal region of human PRKAG1.
- Purification:Protein G purification
- Cat. No.:10679-774
- Supplier no.:PAB4579
Rabbit polyclonal antibody raised against synthetic peptide of PRKAG1.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100
Type: Primary
Antigen: PRKAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse