Anti-Chondroitin-4-Sulfate Mouse Monoclonal Antibody [clone: BE-123]
Catalog # 10703-994
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Chondroitin-4-Sulfate
- Antigen Symbol:Chondroitin-4-Sulfate
- Clonality:Monoclonal
- Clone:BE-123
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Sheep,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Form:Ascites
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from mouse proteoglycan.
- Tested Applications:Radio Immunoassay
- Cat. No.:10703-994
- Supplier no.:MAB6611
Specifications
About this item
Mouse monoclonal antibody raised against Chondroitin-4-Sulfate.
Recommended Dilutions: Western Blot: 1:1000-1:4000; ELISA: 1:400-1:3200; Radio Immunoassay: 1:50-1:6400
Type: Primary
Antigen: Chondroitin-4-Sulfate
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: BE-123
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Bovine, Dog, Human, Mouse, Rat, Sheep